Skip to Content
Merck
All Photos(1)

Key Documents

APO456796561

Apollo Scientific Ltd

Ethyl 2-methyl-4,4,4-trifluoroacetoacetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
198.14
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

47

density

1.258

Iupac Name

ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate

Inchi Code

1S/C7H9F3O3/c1-3-13-6(12)4(2)5(11)7(8,9)10/h4H,3H2,1-2H3

InChI key

YLRGPBKEZVHOAW-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/ethyl-2-methyl-444-trifluoroacetoacetate

Synonym

Ethyl 2-methyl-3-oxo-4,4,4-trifluorobutanoate, Ethyl 2-methyl-3-oxo-4,4,4-trifluorobutyrate

Flash Point

144-146

Melting Point

47

density

1.258

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/ethyl-2-methyl-444-trifluoroacetoacetate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service