Skip to Content
Merck
All Photos(1)

Key Documents

698040

Sigma-Aldrich

2-Bromophenylboronic acid MIDA ester

95%

Synonym(s):

2-(2-Bromophenyl)-6-methyl-1,3,6,2-dioxazaboracane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BBrNO4
CAS Number:
Molecular Weight:
311.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

205-213 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccccc2Br

InChI

1S/C11H11BBrNO4/c1-14-6-10(15)17-12(18-11(16)7-14)8-4-2-3-5-9(8)13/h2-5H,6-7H2,1H3

InChI key

OKCCXZGVZLOVRX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service