Skip to Content
Merck
All Photos(1)

Documents

376167

Sigma-Aldrich

N-(Trimethylsilylmethyl)phthalimide

98%

Synonym(s):

Phthalimidomethyl-trimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO2Si
CAS Number:
Molecular Weight:
233.34
Beilstein:
153462
MDL number:
UNSPSC Code:
12352103

Assay

98%

bp

76-77 °C/0.01 mmHg (lit.)

mp

25-27 °C (lit.)

SMILES string

C[Si](C)(C)CN1C(=O)c2ccccc2C1=O

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service