MilliSentials™ Essential Lab Equipment for Productive, Efficient Labs
Work Smarter
Many protocols require essential lab equipment to tackle routine, repetitive tasks in the laboratory. Unfortunately, current offerings for this equipment are characterized by decades old designs lacking any recent innovation. We are proud to introduce the MilliSentials™ line of general lab equipment aimed at increasing lab efficiency through updated, innovative general lab equipment and devices. Featuring modernized, enhanced devices that simplify routine tasks in the laboratory, these products help streamline your applications.
Section Overiew
View this video for a brief overview on the capabilities of the MilliSentials™ Lab Labeling System.
Adhering to your Highest Standards
The MilliSentials™ Lab Labeling System provides a complete laboratory labeling solution with laboratory grade labels, a compact WIFI capable printer, and custom-designed laboratory labeling software.
- Laboratory grade labels that withstand temperatures from -196 °C (liquid nitrogen) to 100 °C
- Chemical resistance to water, ethanol, IPA, DMSO, methanol, acetonitrile, bleach, acetone, xylene, and toluene
- Adaptable labels that accommodate multiple vessel types and sizes
- Scannable 1D and 2D barcode generation to further optimize sample tracking
- Software custom-built specifically for laboratories, with pre-defined labels for laboratory vessels
- Quick and easy template creation, saving, and sharing of commonly used labels
- Intuitive, easy-to-use thermal transfer printer with hardwire and wireless connection options
- Convenient printing using a laptop, tablet, or mobile device
Enhance your Liquid Handling
The MilliSentials™ Aliquoting Pipette Controller combines the automatic aliquoting function of a repeat pipettor with the basic functions of a standard pipette controller in a single device.
- Built in aliquoting capability automatically dispenses accurate aliquots between 0.5-5.0 mL at the touch of a button
- Touch sensitive aspirate and dispense buttons allow precise control
- Ergonomic design reduces hand strain
Modernize your Vortexer
The MilliSentials™ Vortexer provides powerful agitation to liquid samples within sealed containers in a compact modernized design. The compact and powerful design will accommodate tubes with volumes from 0.2 to 50 mL, vials with volumes from 2.0 to 10 mL, and square bottles up to 150 mL.
- Powerful agitation at a reasonable price provides a valuable option
- Modernized compact design minimizes loss of bench space