Supelco® Carbon Adsorbent Selection Guide
In addition to the tables below that list general guidelines, physical characteristics of Supelco® carbon adsorbents and adsorbent cross-reference, view the report A Tool for Selecting an Adsorbent for Thermal Desorption Applications, which highlights the relative differences among the most commonly used adsorbents with the help of color-coded performance charts.
General Guidelines for Choosing Supelco® Carbon Adsorbents
For multi-bed tubes, use the weaker adsorbent in front of the stronger adsorbent. For example, use Carbopack™ C adsorbent in front of Carbopack™ B adsorbent.
Physical Characteristics of Supelco® Carbon Adsorbents |
---|
Adsorbent Cross Reference |
---|