Merck
Read more about
Cas #: 9004-54-0
Structure
Dextran is a polymer of anhydroglucose.  It is composed of approximately 95% alpha-D-(1-6) linkages. The remaining a(1-3) linkages account for the branching of dextran.1,2,3 Conflicting data on the branch lengths implies that the average branch length is less than three glucose units.4,5  However, other methods indicate branches of greater than 50 glucose units exist.6,7
Native dextran has been found to have a molecular weight (MW) in the range of 9 million to 500 million.8,9,10  Lower MW dextrans will exhibit slightly less branching4 and have a more narrow range of MW distribution.11 Dextrans with MW greater than 10,000 behave as if they are highly branched.  As the MW increases, dextran molecules attain greater symmetry.7,12,13   Dextrans with MW of 2,000 to 10,000 dextran molecules exhibit the properties of an expandable coil.12   At MWs below 2,000 dextran is more rod-like.14
The MW of dextran is measured by one or more of the following methods: low angle laser light scattering15, size exclusion chromatography16,  copper-complexation17, and anthrone reagent18 colorometric reducing-end sugar determination and viscosity12.
Specific Rotation: [α]=+199° 11
Specific Rotation
Figure 1.Specific Rotation
Product Information
We offer dextrans which are derived from Leuconostoc mesenteroides, strain B 512.  Various MWs are produced by limited hydrolysis and fractionation. Our supplier's exact methods are held proprietary. Fractionation can be accomplished by size exclusion chromatography16 or ethanol fractionation in which the largest MW dextrans precipitate first.19
Preparation Instructions
Dextrans are very water soluble, with the exception of the highest MW dextran, D5501 (MW range = 5 million to 40 million).  We test the solubility of dextrans at concentrations generally exceeding 30 mg/mL in water.
Dextrans are also freely soluble in DMSO, formamide, ethylene glycol, and glycerol.11
Neutral-aqueous dextran solutions can be sterilized by autoclaving at 110-115 °C for 30 to 45 minutes.11 Dextran can be hydrolyzed by strong acids at high temperatures. The terminal reducing end group of dextran can be oxidized in alkaline solutions.11
Procedure
Dextran is a high molecular weight, inert, water soluble polymer that has been used in a wide variety of bio-medical applications.
Applications for Unmodified Dextrans:
Plasma Extenders
Dextran solutions have been used as plasma extenders.20
A 10% dextran solution (MW 40,000) exerts a slightly higher colloidal osmotic pressure than plasma proteins.
A 10% dextran (MW 40,000) solution in 0.9% sodium chloride or 5% glucose has reported to be used as a short-term plasma extender for post-operative thrombo-embolic disorders.  After infusion, approximately 70% dextran (MW 40,000) is excreted in the urine unchanged after 24 hours. A small amount is eliminated in the feces.  The remaining dextran is slowly metabolized to glucose.
A 6% solution of Dextran (MW 70,000) exerts a colloidal osmotic pressure similar to that of plasma proteins. Dextrans with MW greater than 50,000 tend to slowly diffuse across the capillary wall and are slowly metabolized to glucose. Approximately 50% of infused dextran MW 70,000 is excreted unchanged in the urine in 24 hours 20.
Centrifugation / Cell and Organelle Separation
Colloidal solutions containing dextran (MW 250,000) have been used to isolate aggregated platelets21 leukocytes22 and lymphocytes23 in blood by centrifugation.
Dextran (MW 40,000) has been used for the isolation of intact nuclei.24
Protein Precipitation
Dextrans have been used to enhance the precipitation and sensitivity of antibody-antigen complexes in immuno-diffusion applications.
Dextran (MW 80,000) was infused into an immunoelectrophoresis gel at a maximum of 80 mg/mL.25
Dextrans (MW 250,000 and 2,000,000) have also been used in similar applications.26
Inhibition of Platelet Aggregation
At lower concentrations Dextrans (MW 10,000-40,000) have been used to inhibit platelet aggregation.27
References
1.
Rankin JC, Jeanes A. 1954. Evaluation of the Periodate Oxidation Method for Structural Analysis of Dextrans. J. Am. Chem. Soc.. 76(17):4435-4441. https://doi.org/10.1021/ja01646a046
2.
Dimler RJ, Wolff IA, Sloan JW, Rist CE. 1955. Interpretation of Periodate Oxidation Data on Degraded Dextran. J. Am. Chem. Soc.. 77(24):6568-6573. https://doi.org/10.1021/ja01629a044
3.
Van Cleve JW, Schaefer WC, Rist CE. 1956. The Structure of NRRL B-512 Dextran. Methylation Studies2. J. Am. Chem. Soc.. 78(17):4435-4438. https://doi.org/10.1021/ja01598a064
4.
Lindberg B, Svensson S, Sjövall J, Zaidi NA. 1968. Structural Studies on Dextran from Leuconostoc mesenteroides NRRL B-512.. Acta Chem. Scand.. 221907-1912. https://doi.org/10.3891/acta.chem.scand.22-1907
5.
Larm O, Lindberg B, Svensson S. 1971. Studies on the length of the side chains of the dextran elaborated by Leuconostoc mesenteroides NRRL B-512. Carbohydrate Research. 20(1):39-48. https://doi.org/10.1016/s0008-6215(00)84947-2
6.
Bovey FA. 1959. Enzymatic polymerization. I. Molecular weight and branching during the formation of dextran. J. Polym. Sci.. 35(128):167-182. https://doi.org/10.1002/pol.1959.1203512813
7.
Senti FR, Hellman NN, Ludwig NH, Babcock GE, Tobin R, Glass CA, Lamberts BL. 1955. Viscosity, sedimentation, and light-scattering properties of fraction of an acid-hydrolyzed dextran. J. Polym. Sci.. 17(86):527-546. https://doi.org/10.1002/pol.1955.120178605
8.
Arond LH, Frank HP. 1954. Molecular Weight Distribution and Molecular Size of a Native Dextran. J. Phys. Chem.. 58(11):953-957. https://doi.org/10.1021/j150521a006
9.
Elias VH. 1959. Ultrazentrifugen- und Diffusionsmessungen an nicht-Newtonschen Lösungen nativer Dextrane. Über extrem große Makromoleküle. IV. Makromol. Chem.. 33(1):166-180. https://doi.org/10.1002/macp.1959.020330112
10.
Antonini E, Bellelli L, Bruzzesi MR, Caputo A, Chiancone E, Rossi-Fanelli A. 1964. Studies on dextran and dextran derivatives. I. Properties of native dextran in different solvents. Biopolymers. 2(1):27-34. https://doi.org/10.1002/bip.1964.360020105
11.
Supplier's data..
12.
Granath KA. 1958. Solution properties of branched dextrans. Journal of Colloid Science. 13(4):308-328. https://doi.org/10.1016/0095-8522(58)90041-2
13.
Wales M. 1979. J.Polym. Sci. 66101.
14.
GEKKO K. 1981. Solution Properties of Dextran and Its Ionic Derivatives.415-438. https://doi.org/10.1021/bk-1981-0150.ch029
15.
Gaylord NG, Beaman RG. 1960. Techniques of polymer characterization. P. W. Allen. Academic Press, New York; Butter-worths, London, 1959. xiv + 256 pp. $9.50.. J. Appl. Polym. Sci.. 3(7):128-128. https://doi.org/10.1002/app.1960.070030723
16.
Granath KA, Flodin P. 1961. Makromol. Chem.. 48(1):160-171. https://doi.org/10.1002/macp.1961.020480116
17.
1953. Journal of Research of the National Bureau of Standards. 5081.
18.
Jermyn M. 1975. Increasing the sensitivity of the anthrone method for carbohydrate. Analytical Biochemistry. 68(1):332-335. https://doi.org/10.1016/0003-2697(75)90713-7
19.
Cramér H. 1949. On the factorization of certain probability distributions. Ark. Mat.. 1(1):61-65. https://doi.org/10.1007/bf02590468
20.
LIPMAN AG. 1993. MARTINDALE: ?Martindale - the Extra Pharmacopoeia? (30th ed), edited by J. E. F. Reynolds. 2(2):124-124. https://doi.org/10.1111/j.2042-7174.1993.tb00740.x
21.
Minor A, Burnett L, State J N. 1953. 53547.
22.
Boyum A, Scan J. 1968. Clin. Lab. Invest.. 21(97):31.
23.
Harris R, Ukaejiofo EO. 1970. Tissue Typing Using a Routine One-Step Lymphocyte Separation Procedure. Br J Haematol. 18(2):229-236. https://doi.org/10.1111/j.1365-2141.1970.tb01436.x
24.
HONDA SI, HONGLADAROM T, LATIES GG. 1966. A New Isolation Medium for Plant Organelles. J Exp Bot. 17(3):460-472. https://doi.org/10.1093/jxb/17.3.460
25.
Hellsing K. 1969. Immune reactions in polysaccharide media. The composition of the antigen?antibody complexes in the precipitin reaction. 114(1):141-144. https://doi.org/10.1042/bj1140141
26.
Ceska M. 1969. Human growth hormone-anti-(growth hormone) immunoprecipitates in the presence of dextrans. 111(4):607-608. https://doi.org/10.1042/bj1110607
27.
Gelin L, Ingelman B. 1961. Acta Chir.. Scand. 122(294):