HPLC Analysis of Sertraline Enantiomers on Astec® CYCLOBOND I 2000 RSP
![HPLC Analysis of Sertraline Enantiomers on Astec® CYCLOBOND I 2000 RSP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/843/839/a035f322-2a70-437e-8344-fd350432afe1/800/a035f322-2a70-437e-8344-fd350432afe1.jpg)
CONDITIONS
column
CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] acetonitrile; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null