HPLC Analysis of Norleucine Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of Norleucine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/283/481/3aa7fab7-8e1b-4eff-a9ba-d4010517cebb/800/3aa7fab7-8e1b-4eff-a9ba-d4010517cebb.jpg)
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] ethanol; (80:20, A:B)
flow velocity
1 mL/min
column temp.
25 °C
detector
UV, 210 nm
injection
5 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC