GC Analysis of Blood Alcohols in Human Plasma on SUPELCOWAX 10 after SPME using 60 μm Carbowax® Fiber
![GC Analysis of Blood Alcohols in Human Plasma on SUPELCOWAX 10 after SPME using 60 μm Carbowax® Fiber suitable for GC, application for SPME](/deepweb/assets/sigmaaldrich/product/images/202/608/03327a3a-a76c-4391-ad0c-98497ab394c0/800/03327a3a-a76c-4391-ad0c-98497ab394c0.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
sample/matrix
blood alcohols at concentrations indicated in human plasma
SPME fiber
60 μm Carbowax
extraction
headspace, 50 °C (5 min)
desorption process
220 °C, 2 min
column
SUPELCOWAX 10, 30 m × 0.25 mm I.D., 0.50 μm (24284)
oven
35 °C (2 min), 10 °C/min to 125 °C (1 min)
detector
FID, 200 °C
carrier gas
helium, 1.0 mL/min constant
injection
10:1 split
liner
0.75 mm I.D. SPME liner
Description
Legal Information
Carbowax is a registered trademark of Union Carbide
null