Merck
Photocatalytic function of the B
Dalton transactions (Cambridge, England : 2003) (2017-12-23)
Hui Tian, Hisashi Shimakoshi, Gyurim Park, Sinheui Kim, Youngmin You, Yoshio Hisaeda
ABSTRACT
A visible light induced three-component catalytic system with the cobalamin derivative (B