Merck
  • Combined shRNA over CRISPR/cas9 as a methodology to detect off-target effects and a potential compensatory mechanism.
Combined shRNA over CRISPR/cas9 as a methodology to detect off-target effects and a potential compensatory mechanism.
Scientific reports (2018-01-10)
Liat Peretz, Elazar Besser, Renana Hajbi, Natania Casden, Dan Ziv, Nechama Kronenberg, Liat Ben Gigi, Sahar Sweetat, Saleh Khawaled, Rami Aqeilan, Oded Behar
ABSTRACT
Inhibition of genes is a powerful approach to study their function. While RNA interference is a widely used method to achieve this goal, mounting evidence indicates that such an approach is prone to off-target effects. An alternative approach to gene function inhibition is genetic mutation, such as the CRISPR/cas9 method. A recent report, however, demonstrated that genetic mutation and inhibition of gene expression do not always give corresponding results. This can be explained by off-target effects, but it was recently shown, at least in one case, that these differences are the result of a compensatory mechanism induced only by genetic mutation. We present here a combination of RNA inhibition and CRISPR/cas9 methods to identify possible off targets as well as potential compensatory effects. This approach is demonstrated by testing a possible role for Sema4B in glioma biology, in which our results implicate Sema4B as having a critical function. In stark contrast, by using shRNA over CRISPR/cas9 combined methodology, we clearly demonstrate that the Sema4B targeted shRNA effects on cell proliferation is the result of off-target effects. Nevertheless, it also revealed that certain splice variants of Sema4B are important for the ability of glioma cells to grow as individual clones.
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)