Merck
Palladium-catalyzed distannylation of ortho-quinodimethanes.
Organic letters (2006-08-25)
Hiroto Yoshida, Saori Nakano, Yasuhito Yamaryo, Joji Ohshita, Atsutaka Kunai
ABSTRACT
An exo-diene moiety of various ortho-quinodimethanes, regardless of its transient character, was inserted into a Sn-Sn sigma-bond of hexabutyldistannane in the presence of a palladium catalyst, giving alpha,alpha'-bis(tributylstannyl)-o-xylenes straightforwardly.
Sigma-Aldrich
Diphenyl-2-pyridylphosphine, 97%