Merck
  • The dynamic recruitment of TRBP to neuronal membranes mediates dendritogenesis during development.
The dynamic recruitment of TRBP to neuronal membranes mediates dendritogenesis during development.
EMBO reports (2017-12-22)
Anna Antoniou, Sharof Khudayberdiev, Agata Idziak, Silvia Bicker, Ralf Jacob, Gerhard Schratt
ABSTRACT
MicroRNAs are important regulators of local protein synthesis during neuronal development. We investigated the dynamic regulation of microRNA production and found that the majority of the microRNA-generating complex, consisting of Dicer, TRBP, and PACT, specifically associates with intracellular membranes in developing neurons. Stimulation with brain-derived neurotrophic factor (BDNF), which promotes dendritogenesis, caused the redistribution of TRBP from the endoplasmic reticulum into the cytoplasm, and its dissociation from Dicer, in a Ca2+-dependent manner. As a result, the processing of a subset of neuronal precursor microRNAs, among them the dendritically localized pre-miR16, was impaired. Decreased production of miR-16-5p, which targeted the BDNF mRNA itself, was rescued by expression of a membrane-targeted TRBP Moreover, miR-16-5p or membrane-targeted TRBP expression blocked BDNF-induced dendritogenesis, demonstrating the importance of neuronal TRBP dynamics for activity-dependent neuronal development. We propose that neurons employ specialized mechanisms to modulate local gene expression in dendrites, via the dynamic regulation of microRNA biogenesis factors at intracellular membranes of the endoplasmic reticulum, which in turn is crucial for neuronal dendrite complexity and therefore neuronal circuit formation and function.
Sigma-Aldrich
Anti-Dicer antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Rabbit Anti-Goat IgG, H & L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Anti-Myosin Va (LF-18) antibody produced in rabbit, ~0.4 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Calnexin Antibody, from rabbit, purified by affinity chromatography