- Combined replacement effects of human modified β-hexosaminidase B and GM2 activator protein on GM2 gangliosidoses fibroblasts.
Combined replacement effects of human modified β-hexosaminidase B and GM2 activator protein on GM2 gangliosidoses fibroblasts.
Biochemistry and biophysics reports (2016-06-08)
Keisuke Kitakaze, Chikako Tasaki, Youichi Tajima, Takatsugu Hirokawa, Daisuke Tsuji, Hitoshi Sakuraba, Kohji Itoh
PMID28955902
ABSTRACT
GM2 gangliosidoses are autosomal recessive lysosomal storage diseases (LSDs) caused by mutations in the
Sigma-Aldrich
Anti-GM2A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution