Merck
S-1: a promising new oral fluoropyrimidine derivative.
Expert opinion on investigational drugs (2009-02-27)
Muhammad Wasif Saif, Kostas N Syrigos, Nikos A Katirtzoglou
ABSTRACT
S-1 is an oral fluoropyrimidine that is designed to improve the antitumor activity of 5-fluorouracil (5-FU) concomitantly with an intent to reduce its toxicity. S-1 consists of tegafur, a prodrug of 5-FU combined with two 5-FU biochemical modulators:5-chloro-2,4-dihydroxypyridine (gimeracil or CDHP), a competitive inhibitor of dihydropyrimidine dehydrogenase and oteracil potassium which inhibits phosphorylation of 5-FU in the gastrointestinal tract decreasing serious gastrointestinal toxicities,including nausea, vomiting, stomatitis and diarrhea. Being an oral agent, S-1 offers convenience of administration and prevents complications of central venous access such as infection, thrombosis and bleeding. S-1 has shown efficacy in both gastrointestinal as well non-gastrointestinal malignancies. The authors review the current literature and provide their expert opinion on the incorporation of S-1 in the treatment of solid malignancies [corrected].
Sigma-Aldrich
Tegafur, ≥98% (HPLC), powder