Merck
  • The orphan nuclear receptor TR4 regulates erythroid cell proliferation and maturation.
The orphan nuclear receptor TR4 regulates erythroid cell proliferation and maturation.
Blood (2017-10-12)
Mary P Lee, Osamu Tanabe, Lihong Shi, Natee Jearawiriyapaisarn, Daniel Lucas, James Douglas Engel
ABSTRACT
The orphan nuclear receptors TR4 (NR2C2) and TR2 (NR2C1) are the DNA-binding subunits of the macromolecular complex, direct repeat erythroid-definitive, which has been shown to repress