Merck
  • Leishmania braziliensis SCD6 and RBP42 proteins, two factors with RNA binding capacity.
Leishmania braziliensis SCD6 and RBP42 proteins, two factors with RNA binding capacity.
Parasites & vectors (2017-12-21)
Paola A Nocua, Cesar A Ramirez, José M Requena, Concepción J Puerta
ABSTRACT
The study of RNA binding proteins (RBPs) is of great relevance for understanding processes like post-transcriptional control of gene expression. The post-transcriptional mechanisms are particularly important in Leishmania parasites and related trypanosomatids since transcriptional regulation is almost absent in them. Thus, RBPs should be essential during the development of these parasites and for survival strategies against the adverse conditions that they face during their life-cycle. This work was aimed to do a structural and biochemical characterization of two Leishmania braziliensis proteins, which were previously found in pull-down assays using an HSP70 RNA as bait. At that time, these proteins were annotated as hypothetical proteins (LbrM.25.2210 and LbrM.30.3080) in the GeneDB database. Structural analysis indicated that these two proteins belong to evolutionarily conserved families; thus, they have been renamed accordingly as LbSCD6 (LbrM.25.2210) and LbRBP42 (LbrM.30.3080). We have demonstrated experimentally that these proteins are RBPs, in agreement with their structural features. Both proteins were able to bind to the complete 3' UTR-II region of HSP70-type II mRNA, and to an A + U rich element (ARE) present in that UTR. Cellular localization assays suggested that both proteins are mainly distributed in the cytoplasm of promastigotes growing at 26 °C, but they accumulate in foci around the nucleus when the parasites are under heat-shock conditions. Also, our study showed that steady-state levels of LbSCD6 and LbRBP42 transcripts decreased significantly during incubation of L. braziliensis promastigotes at heat-shock temperatures. However, in these conditions, the cellular content of both proteins remained unaltered. Our data suggest that LbSCD6 and LbRBP42, as occurs for their orthologues in other organisms, are involved in mRNA regulation, and probably they have a relevant role facing the stress conditions that L. braziliensis encounters during insect-to-mammalian transmission.
Sigma-Aldrich
6-Biopterin, ≥97%