Merck
The general utility of a glycerophosphate-Tris buffered medium.
The Journal of applied bacteriology (1984-04-01)
J Douglas, J Dhillon, S E Smith
ABSTRACT
An easily prepared medium, originally designed for the cultivation of lactic phages, has been found to have much wider application. Experience in its use over a ten year period with a range of physiologically diverse bacteria, for teaching and research are summarized and evaluated.
Sigma-Aldrich
Ī²-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
Ī²-Glycerophosphate disodium salt hydrate, ā‰¤1.0 mol % L-Ī±-isomer