Merck
  • Effect of liraglutide on dietary lipid-induced insulin resistance in humans.
Effect of liraglutide on dietary lipid-induced insulin resistance in humans.
Diabetes, obesity & metabolism (2017-06-13)
Juraj Koska, Lizette Lopez, Karen D'Souza, Tracy Osredkar, James Deer, Julie Kurtz, Arline D Salbe, Sherman M Harman, Peter D Reaven
ABSTRACT
To test whether liraglutide suppresses postprandial elevations in lipids and thus protects against high saturated fatty acid (SFA) diet-induced insulin resistance. In a randomized placebo-controlled crossover study, 32 participants with normal or mildly impaired glucose tolerance received liraglutide and placebo for 3 weeks each. Insulin suppression tests (IST) were conducted at baseline and after a 24-hour SFA-enriched diet after each treatment. Plasma glucose, insulin, triglycerides and non-esterified fatty acids (NEFA) were measured over the initial 8 hours (breakfast and lunch) on the SFA diet. A subset of participants underwent ex vivo measurements of insulin-mediated vasodilation of adipose tissue arterioles and glucose metabolism regulatory proteins in skeletal muscle. Liraglutide reduced plasma glucose, triglycerides and NEFA concentrations during the SFA diet (by 50%, 25% and 9%, respectively), and the SFA diet increased plasma glucose during the IST (by 36%; all P < .01 vs placebo). The SFA diet-induced impairment of vasodilation on placebo (-9.4% vs baseline; P < .01) was ameliorated by liraglutide (-4.8%; P = .1 vs baseline). In skeletal muscle, liraglutide abolished the SFA-induced increase in thioredoxin-interacting protein (TxNIP) expression (75% decrease; P < .01 vs placebo) and increased 5'AMP-activated protein kinase (AMPK) phosphorylation (50% vs -3%; P = .04 vs placebo). Liraglutide blunted the SFA-enriched diet-induced peripheral insulin resistance. This effect may be related to improved microvascular function and modulation of TxNIP and AMPK pathways in skeletal muscle.
Product Description
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O