Merck
  • High efficiency of dye-sensitized solar cells based on metal-free indoline dyes.
High efficiency of dye-sensitized solar cells based on metal-free indoline dyes.
Journal of the American Chemical Society (2004-09-30)
Tamotsu Horiuchi, Hidetoshi Miura, Kouichi Sumioka, Satoshi Uchida
ABSTRACT
We now report metal-free organic dyes having a new type of indoline structure, which exhibits high efficiencies in dye-sensitized solar cells. The solar energy to current conversion efficiencies with the new indoline dye was 6.51%. Under the same conditions, the N3 dye was 7.89% and the N719 dye was 8.26%. The new indoline dye was optimized for the amount of 4-tert-butyl pyridine in the electrolyte and cholic acid as a coadsorbent. Subsequently, the solar energy to current conversion efficiencies reached 8.00%. This value was the highest obtained efficiency for dye-sensitized solar cells based on metal-free organic dyes without an antireflection layer.
Sigma-Aldrich
D149 Dye, 98% (HPLC)