Merck
  • RECQ-like helicases Sgs1 and BLM regulate R-loop-associated genome instability.
RECQ-like helicases Sgs1 and BLM regulate R-loop-associated genome instability.
The Journal of cell biology (2017-10-19)
Emily Yun-Chia Chang, Carolina A Novoa, Maria J Aristizabal, Yan Coulombe, Romulo Segovia, Richa Chaturvedi, Yaoqing Shen, Christelle Keong, Annie S Tam, Steven J M Jones, Jean-Yves Masson, Michael S Kobor, Peter C Stirling
ABSTRACT
Sgs1, the orthologue of human Bloom's syndrome helicase BLM, is a yeast DNA helicase functioning in DNA replication and repair. We show that SGS1 loss increases R-loop accumulation and sensitizes cells to transcription-replication collisions. Yeast lacking SGS1 accumulate R-loops and γ-H2A at sites of Sgs1 binding, replication pausing regions, and long genes. The mutation signature of sgs1Δ reveals copy number changes flanked by repetitive regions with high R-loop-forming potential. Analysis of BLM in Bloom's syndrome fibroblasts or by depletion of BLM from human cancer cells confirms a role for Sgs1/BLM in suppressing R-loop-associated genome instability across species. In support of a potential direct effect, BLM is found physically proximal to DNA:RNA hybrids in human cells, and can efficiently unwind R-loops in vitro. Together, our data describe a conserved role for Sgs1/BLM in R-loop suppression and support an increasingly broad view of DNA repair and replication fork stabilizing proteins as modulators of R-loop-mediated genome instability.
Sigma-Aldrich
Rabbit anti-BLM Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)