Merck
  • 14-3-3ζ loss leads to neonatal lethality by microRNA-126 downregulation-mediated developmental defects in lung vasculature.
14-3-3ζ loss leads to neonatal lethality by microRNA-126 downregulation-mediated developmental defects in lung vasculature.
Cell & bioscience (2017-11-10)
Jun Yang, Sonali Joshi, Qingfei Wang, Ping Li, Hai Wang, Yan Xiong, Yi Xiao, Jinyang Wang, Jan Parker-Thornburg, Richard R Behringer, Dihua Yu
ABSTRACT
The 14-3-3 family of proteins have been reported to play an important role in development in various mouse models, but the context specific developmental functions of 14-3-3ζ remain to be determined. In this study, we identified a context specific developmental function of 14-3-3ζ. Targeted deletion of 14-3-3ζ in the C57Bl/6J murine genetic background led to neonatal lethality due to respiratory distress and could be rescued by out-breeding to the CD-1 or backcrossing to the FVB/NJ congenic background. Histological analysis of lung sections from 18.5 days post coitum embryos (dpc) showed that 14-3-3ζ-/- lung development is arrested at the pseudoglandular stage and exhibits vascular defects. The expression of miR-126, an endothelial-specific miRNA known to regulate lung vascular integrity was down-regulated in the lungs of the 14-3-3ζ-/- embryos in the C57Bl/6J background as compared to their wild-type counterparts. Loss of 14-3-3ζ in endothelial cells inhibited the angiogenic capability of the endothelial cells as determined by both trans-well migration assays and tube formation assays and these defects could be rescued by re-expressing miR-126. Mechanistically, loss of 14-3-3ζ led to reduced Erk1/2 phosphorylation resulting in attenuated binding of the transcription factor Ets2 on the miR-126 promoter which ultimately reduced expression of miR-126. Our data demonstrates that miR-126 is an important angiogenesis regulator that functions downstream of 14-3-3ζ and downregulation of miR-126 plays a critical role in 14-3-3ζ-loss induced defects in lung vasculature in the C57Bl/6J genetic background.
Sigma-Aldrich
Anti-Aquaporin 5 Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid