Merck
Genome reprogramming in
Physiological genomics (2017-09-10)
Ceyhun Bereketoglu, Kazim Yalcin Arga, Serpil Eraslan, Bulent Mertoglu
ABSTRACT
Bioaccumulative environmental estrogen, nonylphenol (NP; 4-nonylphenol), is widely used as a nonionic surfactant and can affect human health. Since genomes of
Sigma-Aldrich
Pyrimidine, ≥98.0%
Pricing and availability is not currently available.