Merck
  • Effects of different sterilization methods on the physico-chemical and bioresponsive properties of plasma-treated polycaprolactone films.
Effects of different sterilization methods on the physico-chemical and bioresponsive properties of plasma-treated polycaprolactone films.
Biomedical materials (Bristol, England) (2017-01-25)
Rouba Ghobeira, Charlot Philips, Heidi Declercq, Pieter Cools, Nathalie De Geyter, Ria Cornelissen, Rino Morent
ABSTRACT
For most tissue engineering applications, surface modification and sterilization of polymers are critical aspects determining the implant success. The first part of this study is thus dedicated to modifying polycaprolactone (PCL) surfaces via plasma treatment using a medium pressure dielectric barrier discharge, while the second part focuses on the sterilization of plasma-modified PCL. Chemical and physical surface changes are examined making use of water contact angle goniometry (WCA), x-ray photoelectron spectroscopy and atomic force microscopy. Bioresponsive properties are evaluated by performing cell culture tests. The results show that air and argon plasmas decrease the WCA significantly due to the incorporation of oxygen-containing functionalities onto the PCL surface, without modifying its morphology. Extended treatment times lead to PCL degradation, especially in the case of air plasma. In addition to surface modification, the plasma potential to sterilize PCL is studied with appropriate treatment times, but sterility has not been achieved so far. Therefore, plasma-modified films are subjected to UV, H
Product Description
Millipore
Tryptic Soy Broth - Dehydrated Culture Media, NutriSelect® Basic, powder, suitable for microbiology