Merck
  • Engineering thiophene-based nanoparticles to induce phototransduction in live cells under illumination.
Engineering thiophene-based nanoparticles to induce phototransduction in live cells under illumination.
Nanoscale (2017-06-27)
M Zangoli, F Di Maria, E Zucchetti, C Bossio, M R Antognazza, G Lanzani, R Mazzaro, F Corticelli, M Baroncini, G Barbarella
ABSTRACT
We report that nanoparticles prepared from appropriately functionalized polythiophenes once administered to live cells can acquire phototransduction properties under illumination, becoming photoactive sites able to absorb visible light and convert it to an electrical signal through cell membrane polarization. Amine-reactive fluorescent nanoparticles with pendant N-succinimidyl-ester groups (NPs-NHS) are prepared from polythiophenes alternating unsubstituted and 3-(2,5-dioxopyrrolidin-1-yl-8-octanoate)-substituted thiophenes by a nanoprecipitation method. By
Sigma-Aldrich
2,2′-Bithiophene, 99%
Sigma-Aldrich
3-Bromothiophene, 97%