Merck
  • How does secondary processing affect the physicochemical properties of inhalable salbutamol sulphate particles? A temporal investigation.
How does secondary processing affect the physicochemical properties of inhalable salbutamol sulphate particles? A temporal investigation.
International journal of pharmaceutics (2017-06-18)
Joana T Pinto, Snezana Radivojev, Sarah Zellnitz, Eva Roblegg, Amrit Paudel
ABSTRACT
As pulmonary drug delivery is extended from low doses to high doses, physicochemical characteristics of the active pharmaceutical ingredient gain importance in the development of dry powder inhalers. Therefore, the present work aims to understand the impact of distinct engineering techniques on the process induced physicochemical characteristics of salbutamol sulphate particles over time. The particle engineering techniques chosen were jet-milling and spray-drying, two well used processes in the production of predominately crystalline and amorphous inhalable particles, respectively. Fourier transform infrared spectroscopy, modulated differential scanning calorimetry, particle size distribution and tensiometry experiments were used to characterise the engineered powders immediately, 7, 14 and 21 days after production. The rugged spherical amorphous particles (3.75±0.08μm) obtained via spray-drying showed that they were capable of forming strong agglomerates (5.01±0.22μm) through "amorphous bridging". On the other hand, jet-milling produced smaller (2.06±0.08μm), crystalline, irregular shaped particles with a very large surface area (11.04±0.10m
Sigma-Aldrich
Sodium hexanesulfonate, ≥98% (elemental analysis)