Merck
  • Purification and In Situ Ligand Exchange of Metal-Carboxylate-Treated Fluorescent InP Quantum Dots via Gel Permeation Chromatography.
Purification and In Situ Ligand Exchange of Metal-Carboxylate-Treated Fluorescent InP Quantum Dots via Gel Permeation Chromatography.
The journal of physical chemistry letters (2017-08-12)
Adam Roberge, Jennifer L Stein, Yi Shen, Brandi M Cossairt, Andrew B Greytak
ABSTRACT
Recently the addition of M
Sigma-Aldrich
Hexylphosphonic acid, 95%
Sigma-Aldrich
Ferrocene, 98%