Merck
  • Clinical outcomes following unilateral adrenalectomy in patients with primary aldosteronism.
Clinical outcomes following unilateral adrenalectomy in patients with primary aldosteronism.
QJM : monthly journal of the Association of Physicians (2017-02-10)
M J Hannon, W C Sze, R Carpenter, L Parvanta, M Matson, A Sahdev, M R Druce, D M Berney, M Waterhouse, S A Akker, W M Drake
ABSTRACT
In approximately half of cases of primary aldosteronism (PA), the cause is a surgically-resectable unilateral aldosterone-producing adrenal adenoma. However, long-term data on surgical outcomes are sparse. We report on clinical outcomes post-adrenalectomy in a cohort of patients with PA who underwent surgery. Retrospective review of patients treated for PA in a single UK tertiary centre. Of 120 consecutive patients investigated for PA, 52 (30 male, median age 54, range 30-74) underwent unilateral complete adrenalectomy. Blood pressure, number of antihypertensive medications, and serum potassium were recorded before adrenalectomy, and after a median follow-up period of 50 months (range 7-115). Recumbent renin and aldosterone were measured, in the absence of interfering antihypertensive medication, ≥3months after surgery, to determine if PA had been biochemically cured. Overall, blood pressure improved from a median (range) 160/95 mmHg (120/80-250/150) pre-operatively to 130/80 mmHg (110/70-160/93), P < 0.0001. 24/52 patients (46.2%) had cured hypertension, with a normal blood pressure post-operatively on no medication. 26/52 (50%) had improved hypertension. 2/52 patients (3.8%) showed no improvement in blood pressure post-operatively. Median (range) serum potassium level increased from 3.2 (2.3-4.7) mmol/l pre-operatively to 4.4 mmol/l (3.3-5.3) post-operatively, P < 0.0001). Median (range) number of antihypertensive medications used fell from 3 (0-6) pre- to 1 post-operatively (range 0-4), P < 0.0001. Unilateral adrenalectomy provides excellent long-term improvements in blood pressure control, polypharmacy and hypokalaemia in patients with lateralizing PA. These data may help inform discussions with patients contemplating surgery.
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)