MilliporeSigma
  • Comparison of air samplers for determination of isocyanic acid and applicability for work environment exposure assessment.
Comparison of air samplers for determination of isocyanic acid and applicability for work environment exposure assessment.
Environmental science. Processes & impacts (2017-08-02)
Mikolaj Jan Jankowski, Raymond Olsen, Yngvar Thomassen, Paal Molander
ABSTRACT
Isocyanic acid (ICA) is one of the most abundant isocyanates formed during thermal decomposition of polyurethane (PUR), and other nitrogen containing polymers. Hot-work, such as flame cutting, forging, grinding, turning and welding may give rise to thermal decomposition of said polymers potentially forming significant amounts of ICA. A newly launched dry denuder sampler for airborne isocyanates using di-n-butylamine (DBA) demonstrated build-up of background ICA-DBA over time. Build-up of background ICA-DBA was not observed when stored at inert conditions (Ar atmosphere) for 84 days. Thus, freshly prepared denuders were used. The sampling efficiency of ICA using freshly prepared denuder samplers (0.2 L min
Sigma-Aldrich
Ethyl isocyanate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-(2-Methoxyphenyl)piperazine, 98%