Merck
  • Metabolic profiling of tyrosine, tryptophan, and glutamate in human urine using gas chromatography-tandem mass spectrometry combined with single SPE cleanup.
Metabolic profiling of tyrosine, tryptophan, and glutamate in human urine using gas chromatography-tandem mass spectrometry combined with single SPE cleanup.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-03-25)
Hyun Ju Shin, Na Hyun Park, Wonwoong Lee, Man Ho Choi, Bong Chul Chung, Jongki Hong
ABSTRACT
The tyrosine, tryptophan, and glutamate metabolic pathways play key roles on pathological state of neuronal functions and the change of their levels in biological systems reflects the progress degree of neuronal diseases. Comprehensive profiling of these metabolites is important to find new biomarkers for diagnosis or prognosis of various neuronal diseases. However, the overall profiling analysis of various neurochemicals in biological sample is confronted with several limitations due to their low concentration and physicochemical properties and the coexistence of matrices. We developed an efficient and feasible method using gas chromatography-tandem mass spectrometry (GC-MS/MS). Wide-bore mixed cation exchange (MCX) SPE process enables a rapid and effective cleanup of 20 neurochemicals even including acidic and basic neurochemicals in a single SPE cartridge by using different composition of eluents. Selective derivatization of various types of metabolites was applied to achieve highly chromatographic separation and sensitive mass detection. Appropriate selection of precursor and product transition ions used in multiple reaction-monitoring (MRM) mode based on the MS/MS fragmentations of the derivatized neurochemicals could be significantly minimized the matrix effects and enhanced the reliability of quantification results. The developed method was validated in terms of linearity, limits of detection, precision, accuracy, and matrix effects. The intra- and inter-assay analytical variations were less than 10%. The overall linearity for all of the targets was excellent (R
Sigma-Aldrich
4-Aminobutyric acid-2,2,3,3,4,4-d6, 97 atom % D
Sigma-Aldrich
5-Hydroxyindole-3-acetic acid, ≥98% (HPLC), crystalline