- Effects of bacterial direct-fed microbials on ruminal characteristics, methane emission, and milk fatty acid composition in cows fed high- or low-starch diets.
Effects of bacterial direct-fed microbials on ruminal characteristics, methane emission, and milk fatty acid composition in cows fed high- or low-starch diets.
Journal of dairy science (2017-02-06)
C Philippeau, A Lettat, C Martin, M Silberberg, D P Morgavi, A Ferlay, C Berger, P Nozière
PMID28161181
ABSTRACT
This study investigated the effects of bacterial direct-fed microbials (DFM) on ruminal fermentation and microbial characteristics, methane (CH