Merck
  • Reductive dechlorination of trichloroethylene by polyvinylpyrrolidone stabilized nanoscale zerovalent iron particles with Ni.
Reductive dechlorination of trichloroethylene by polyvinylpyrrolidone stabilized nanoscale zerovalent iron particles with Ni.
Journal of hazardous materials (2017-07-25)
Macharla Arun Kumar, Sungjun Bae, Seunghee Han, Yoonseok Chang, Woojin Lee
ABSTRACT
We developed a novel stabilized nanoscale zerovalent iron (NZVI) particles with Ni using an electron conducting polymer, polyvinylpyrrolidone (PVP), to selectively dechlorinate trichloroethylene (TCE) to non-toxic intermediates. The size of the PVP stabilized NZVI-Ni ((PVP-NZVI-Ni), average diameter: ∼20nm) is smaller than that of bare NZVI (50-80nm) due to the prevention of agglomeration of the resultant iron particles by PVP. PVP-NZVI-Ni showed a complete removal of TCE in 1h with superior dechlorination kinetics (k
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, ≥97%