Merck
  • A New Triazine-Based Covalent Organic Framework for High-Performance Capacitive Energy Storage.
A New Triazine-Based Covalent Organic Framework for High-Performance Capacitive Energy Storage.
ChemSusChem (2017-01-07)
Piyali Bhanja, Kousik Bhunia, Sabuj K Das, Debabrata Pradhan, Ryuto Kimura, Yuh Hijikata, Stephan Irle, Asim Bhaumik
ABSTRACT
The new covalent organic framework material TDFP-1 was prepared through a solvothermal Schiff base condensation reaction of the monomers 1,3,5-tris-(4-aminophenyl)triazine and 2,6-diformyl-4-methylphenol. Owing to its high specific surface area of 651 m
Sigma-Aldrich
4-Aminobenzonitrile, 98%