Merck
  • Visible light assisted hydrogen generation from complete decomposition of hydrous hydrazine using rhodium modified TiO
Visible light assisted hydrogen generation from complete decomposition of hydrous hydrazine using rhodium modified TiO
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2017-05-27)
Pawan Kumar, Anurag Kumar, Clémence Queffélec, Dietrich Gudat, Qi Wang, Suman L Jain, Rabah Boukherroub, Sabine Szunerits
ABSTRACT
Hydrogen is considered to be an ideal energy carrier, which produces only water when combined with oxygen and thus has no detrimental effect on the environment. While the catalytic decomposition of hydrous hydrazine for the production of hydrogen is well explored, little is known about its photocatalytic decomposition. The present paper describes a highly efficient photochemical methodology for the production of hydrogen through the decomposition of aqueous hydrazine using titanium dioxide nanoparticles modified with a Rh(i) coordinated catechol phosphane ligand (TiO
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Allyl methyl sulfone, 96%