Merck
  • Inhibition of glutamate decarboxylase (GAD) by ethyl ketopentenoate (EKP) induces treatment-resistant epileptic seizures in zebrafish.
Inhibition of glutamate decarboxylase (GAD) by ethyl ketopentenoate (EKP) induces treatment-resistant epileptic seizures in zebrafish.
Scientific reports (2017-08-05)
Yifan Zhang, Michiel Vanmeert, Aleksandra Siekierska, Annelii Ny, Jubi John, Geert Callewaert, Eveline Lescrinier, Wim Dehaen, Peter A M de Witte, Rafal M Kaminski
ABSTRACT
Epilepsy is a chronic brain disorder characterized by recurrent seizures due to abnormal, excessive and synchronous neuronal activities in the brain. It affects approximately 65 million people worldwide, one third of which are still estimated to suffer from refractory seizures. Glutamic acid decarboxylase (GAD) that converts glutamate into GABA is a key enzyme in the dynamic regulation of neural network excitability. Importantly, clinical evidence shows that lowered GAD activity is associated with several forms of epilepsy which are often treatment resistant. In the present study, we synthetized and explored the possibility of using ethyl ketopentenoate (EKP), a lipid-permeable GAD-inhibitor, to induce refractory seizures in zebrafish larvae. Our results demonstrate that EKP evoked robust convulsive locomotor activities, excessive epileptiform discharges and upregulated c-fos expression in zebrafish. Moreover, transgenic animals in which neuronal cells express apoaequorin, a Ca
Sigma-Aldrich
Pyrrolidone hydrotribromide, 97%
Sigma-Aldrich
Coelenterazine h, solid