Merck
  • Pentraxin 3 and other inflammatory biomarkers related to atrial fibrillation in cardiac surgery.
Pentraxin 3 and other inflammatory biomarkers related to atrial fibrillation in cardiac surgery.
Perfusion (2016-11-18)
Zdenka Holubcova, Pavel Kunes, Jiri Mandak, Dana Vlaskova, Martina Kolackova, Drahomira Holmannova, Ctirad Andrys, Jan Krejsek, Tomas Holubec
ABSTRACT
The aim was to evaluate the association between perioperative inflammatory biomarkers and atrial fibrillation (AF) in cardiac surgical patients. Forty-two patients undergoing cardiac surgery were divided into three groups according to the occurrence of AF: Group A (n = 22) - patients with no AF, Group B (n = 11) - patients with new onset AF postoperatively and Group C (n = 9) - patients with preoperative history of atrial fibrillation. The serum levels of PTX3, CRP, TLR2, IL-8, IL-18, sFas, MMP-7 and MMP-8 were measured at the following time points: before surgery, immediately and 6 h after surgery and on the 1 Serum levels of PTX3 showed a significant difference between Groups A and C on the 3 This study demonstrates significantly different dynamics of PTX3, IL-8 and sFas levels after cardiac surgery in relation to AF.
Sigma-Aldrich
Mouse Ptx3 / Pentraxin-related Protein PTX3 ELISA Kit
Sigma-Aldrich
Human TLR2 / Toll-like Receptor 2 ELISA Kit