Merck
Association of non-synonymous variants in
Journal of geriatric cardiology : JGC (2016-01-01)
Yuko Maeda, Noriko Sato, Makiko Naka-Mieno, Seijiro Mori, Tomio Arai, Masashi Tanaka, Masaaki Muramatsu, Motoji Sawabe
ABSTRACT
Abdominal aortic aneurysm (AAA) is a multifactorial disease with strong genetic components. Various genetic loci have been associated with clinical AAA, but few studies have investigated pathological AAA, an intermediate phenotype of the disease. We examined 2263 consecutive autopsies of older Japanese subjects from a study on geriatric diseases in Japanese individuals (The JG-SNP study). The presence of AAA was determined with a pathological diagnosis during autopsy. Single nucleotide variants (SNVs) associated with AAA were determined with an Illumina HumanExome Beadchip array. Logistic regression analyses were performed to determine genetic associations. Age, gender, and other risk factors of AAA were analyzed as covariates. 118 subjects with AAA and 2145 subjects without AAA were analyzed in a case-control setting. No variants reached significance after applying the Bonferroni correction ( This study suggests that
Sigma-Aldrich
Anti-WIPF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution