Merck
  • Enantiomeric NMR signal separation behavior and mechanism of samarium(III) and neodymium(III) complexes with (S,S)-ethylenediamine-N,N'-disuccinate.
Enantiomeric NMR signal separation behavior and mechanism of samarium(III) and neodymium(III) complexes with (S,S)-ethylenediamine-N,N'-disuccinate.
Chirality (2017-04-14)
Sen-Ichi Aizawa, Masaru Okano, Takahiro Kidani
ABSTRACT
Enantiomeric
Sigma-Aldrich
Neodymium(III) nitrate hexahydrate, 99.9% trace metals basis