Merck
  • Astrocytic Orosomucoid-2 Modulates Microglial Activation and Neuroinflammation.
Astrocytic Orosomucoid-2 Modulates Microglial Activation and Neuroinflammation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-02-15)
Myungjin Jo, Jong-Heon Kim, Gyun Jee Song, Minchul Seo, Eun Mi Hwang, Kyoungho Suk
ABSTRACT
Orosomucoid (ORM) is an acute-phase protein that belongs to the immunocalin subfamily, a group of small-molecule-binding proteins with immunomodulatory functions. Little is known about the role of ORM proteins in the CNS. The aim of the present study was to investigate the brain expression of ORM and its role in neuroinflammation. Expression of Orm2, but not Orm1 or Orm3, was highly induced in the mouse brain after systemic injection of lipopolysaccharide (LPS). Plasma levels of ORM2 were also significantly higher in patients with cognitive impairment than in normal subjects. RT-PCR, Western blot, and immunofluorescence analyses revealed that astrocytes are the major cellular sources of ORM2 in the inflamed mouse brain. Recombinant ORM2 protein treatment decreased microglial production of proinflammatory mediators and reduced microglia-mediated neurotoxicity
Sigma-Aldrich
MISSION® esiRNA, targeting human ORM2