Merck
  • Synthesis of Ag(I) camphor sulphonylimine complexes and assessment of their cytotoxic properties against cisplatin-resistant A2780cisR and A2780 cell lines.
Synthesis of Ag(I) camphor sulphonylimine complexes and assessment of their cytotoxic properties against cisplatin-resistant A2780cisR and A2780 cell lines.
Journal of inorganic biochemistry (2016-11-12)
João M S Cardoso, Isabel Correia, Adelino M Galvão, Fernanda Marques, M Fernanda N N Carvalho
ABSTRACT
Camphorsulphonylimine complexes [Ag(NO
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, suitable for substrate for DNase (Readily soluble)
Sigma-Aldrich
N,N-Diphenylhydrazine hydrochloride, 97%