Merck
  • Effect of antigastrin (SC-15396) on amylase release and 2-deoxyglucose uptake in mouse pancreatic acini.
Effect of antigastrin (SC-15396) on amylase release and 2-deoxyglucose uptake in mouse pancreatic acini.
Digestion (1985-01-01)
Y Iwamoto, R Nakamura, Y Akanuma, T Kuzuya
ABSTRACT
The effects of antigastrin (SC-15396) on amylase release and 2-deoxyglucose uptake were studied in dispersed acini from mouse pancreas. Antigastrin at concentrations between 0.5 and 2 mM inhibited cholecystokinin (CCK)-stimulated amylase release and 2-deoxyglucose uptake in a dose-dependent fashion. Antigastrin at concentrations between 0.5 and 2 mM inhibited carbachol-stimulated amylase release and 2-deoxyglucose uptake as well. In addition, the drug (2 mM) inhibited bombesin-stimulated amylase release and 2-deoxyglucose uptake. In contrast, the stimulation of amylase release by Ca2+ ionophore A23187 was not inhibited by antigastrin. The results, therefore, suggest that antigastrin nonselectively inhibits the actions of these secretagogues probably at their receptor sites.
Sigma-Aldrich
2-Amino-9H-pyrido[2-3-b]indole, ≥98% (HPLC)