Merck
  • RACK-1, a receptor for activated C kinase, interacts with the transcription factor NFAT and represses its transactivation.
RACK-1, a receptor for activated C kinase, interacts with the transcription factor NFAT and represses its transactivation.
Molecules and cells (2003-01-11)
Juhyun Han, Minjung Kim, Ji-Hyun Oum, Heejoon Myung, Seong-Wook Lee, Sunjoo Jeong, Surendra Sharma, Jungchan Park
ABSTRACT
To isolate and characterize a novel protein that interacts with nuclear factor of activated T cells (NFAT) and potentially regulates its activity, we screened a Jurkat cDNA library by using the NFAT regulatory domain as bait in the yeast two-hybrid system. RACK-1, a receptor for activated protein kinase C and a homologue of the G-protein beta subunit, was identified as a NFAT-binding protein. Mammalian two hybrid tests in CV-1 cells and a coimmunoprecipitation assay confirmed protein-protein interaction between NFAT and RACK-1. In addition, overexpression of RACK-1 specifically suppressed transcriptional activation derived by NFAT, but not by NF-kappaB. These results demonstrate RACK-1 as a potent negative modulator of NFAT activation and suggest a novel mechanism in NFAT regulation.