Merck
Novel
Nuclear medicine and biology (2016-10-25)
Min Liu, Yumin Zheng, Ugur Avcibasi, Shuang Liu
ABSTRACT
In this study, novel Complexes [ Both azide coligand and boronate caps had significant impact on the heart uptake and myocardial retention of complexes [
Sigma-Aldrich
1,2-Cyclohexanedione dioxime, 96%