- Rational design of 6-(2,4-diaminopyrimidinyl)-1,4-benzoxazin-3-ones as small molecule renin inhibitors.
Rational design of 6-(2,4-diaminopyrimidinyl)-1,4-benzoxazin-3-ones as small molecule renin inhibitors.
Bioorganic & medicinal chemistry (2007-06-19)
Noel A Powell, Fred L Ciske, Cuiman Cai, Daniel D Holsworth, Ken Mennen, Chad A Van Huis, Mehran Jalaie, Jacqueline Day, Michelle Mastronardi, Pat McConnell, Igor Mochalkin, Erli Zhang, Michael J Ryan, John Bryant, Wendy Collard, Suzie Ferreira, Chungang Gu, Roxane Collins, Jeremy J Edmunds
PMID17574423
ABSTRACT
We report the design and synthesis of a series of 6-(2,4-diaminopyrimidinyl)-1,4-benzoxazin-3-ones as orally bioavailable small molecule inhibitors of renin. Compounds with a 2-methyl-2-aryl substitution pattern exhibit potent renin inhibition and good permeability, solubility, and metabolic stability. Oral bioavailability was found to be dependent on metabolic clearance and cellular permeability, and was optimized through modulation of the sidechain that binds in the S3(sp) subsite.