Merck
  • Functional 3D Neural Mini-Tissues from Printed Gel-Based Bioink and Human Neural Stem Cells.
Functional 3D Neural Mini-Tissues from Printed Gel-Based Bioink and Human Neural Stem Cells.
Advanced healthcare materials (2016-03-31)
Qi Gu, Eva Tomaskovic-Crook, Rodrigo Lozano, Yu Chen, Robert M Kapsa, Qi Zhou, Gordon G Wallace, Jeremy M Crook
ABSTRACT
Direct-write printing of stem cells within biomaterials presents an opportunity to engineer tissue for in vitro modeling and regenerative medicine. Here, a first example of constructing neural tissue by printing human neural stem cells that are differentiated in situ to functional neurons and supporting neuroglia is reported. The supporting biomaterial incorporates a novel clinically relevant polysaccharide-based bioink comprising alginate, carboxymethyl-chitosan, and agarose. The printed bioink rapidly gels by stable cross-linking to form a porous 3D scaffold encapsulating stem cells for in situ expansion and differentiation. Differentiated neurons form synaptic contacts, establish networks, are spontaneously active, show a bicuculline-induced increased calcium response, and are predominantly gamma-aminobutyric acid expressing. The 3D tissues will facilitate investigation of human neural development, function, and disease, and may be adaptable for engineering other 3D tissues from different stem cell types.
Sigma-Aldrich
ReNcell CX Human Neural Progenitor Cell Line, ReNcell CX is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons & glial cells.