- Glimepiride, a new once-daily sulfonylurea. A double-blind placebo-controlled study of NIDDM patients. Glimepiride Study Group.
Glimepiride, a new once-daily sulfonylurea. A double-blind placebo-controlled study of NIDDM patients. Glimepiride Study Group.
To compare the efficacy and safety of two daily doses of the new sulfonylurea, glimepiride (Amaryl), each as a once-daily dose or in two divided doses, in patients with NIDDM. Of the previously treated NIDDM patients, 416 entered this multicenter randomized double-blind placebo-controlled fixed-dose study. After a 3-week placebo washout, patients received a 14-week course of placebo or glimepiride 8 mg q.d., 4 mg b.i.d., 16 mg q.d., or 8 mg b.i.d. Fasting plasma glucose (FPG) and HbA1c values were similar at baseline in all treatment groups. The placebo group's FPG value increased from 13.0 mmol/l at baseline to 14.5 mmol/l at the last evaluation endpoint (P < or = 0.001). In contrast, FPG values in the four glimepiride groups decreased from a range of 12.4-12.9 mmol/l at baseline to a range of 8.6-9.8 mmol/l at endpoint (P < or = 0.001, within-group change from baseline; P < or = 0.001, between-group change [vs. placebo] from baseline). Two-hour postprandial plasma glucose (PPG) findings were consistent with FPG findings. In the placebo group, the HbA1c value increased from 7.7% at baseline to 9.7% at endpoint (P < or = 0.001), whereas HbA1c values for the glimepiride groups were 7.9-8.1% at baseline and 7.4-7.6% at endpoint (P < or = 0.001, within-group change from baseline; P < or = 0.001, between-group change from baseline). There were no meaningful differences in glycemic variables between daily doses of 8 and 16 mg or between once- and twice-daily dosing. Adverse events and laboratory data demonstrate that glimepiride has a favorable safety profile. Glimepiride is an effective and well-tolerated oral glucose-lowering agent. The results of this study demonstrate maximum effectiveness can be achieved with 8 mg q.d. of glimepiride in NIDDM subjects.