Merck
  • Spleen tyrosine kinase (SYK) is a potential target for the treatment of cutaneous lupus erythematosus patients.
Spleen tyrosine kinase (SYK) is a potential target for the treatment of cutaneous lupus erythematosus patients.
Experimental dermatology (2016-02-26)
Christine Braegelmann, Michael Hölzel, Valerie Ludbrook, Marion Dickson, Nil Turan, Sandra Ferring-Schmitt, Sonja Sternberg, Thomas Bieber, Annegret Kuhn, Joerg Wenzel
ABSTRACT
Spleen tyrosine kinase (SYK) is a protein kinase involved in cell proliferation and the regulation of inflammatory pathways. Due to the increasing evidence that kinase inhibitors have potential as specific anti-inflammatory drugs, we have investigated the potential for SYK inhibition as a therapeutic target in autoimmune diseases, particularly cutaneous lupus erythematosus (CLE). Skin samples of patients with different CLE subtypes and appropriate controls were analysed for the expression of SYK and SYK-associated pro-inflammatory mediators via gene expression analysis and immunohistochemistry. The functional role of SYK in keratinocytes was investigated in vitro, using LE-typical pro-inflammatory stimuli and a selective inhibitor of SYK. SYK-associated genes are strongly upregulated in CLE skin lesions. Importantly, phosphorylated SYK (pSYK) is strongly expressed by several immune cell types and also keratinocytes in CLE skin. In vitro, immunostimulatory nucleic acids are capable of inducing SYK phosphorylation in keratinocytes leading to the induction of pro-inflammatory cytokines, while small-molecule SYK inhibition decreases the expression of these proteins. The results demonstrate that pSYK is expressed by immune cells and keratinocytes in skin lesions of CLE patients. LE-typical stimuli induce the expression of pSYK in vitro. Small-molecule SYK inhibition leads to a reduction of pSYK expression and downregulation of pro-inflammatory cytokines in keratinocytes. We therefore believe that pSYK provides a potential future drug target for the treatment of patients who suffer from CLE and related skin disorders. Specifically, our study reveals evidence supporting the use of topical SYK inhibitors in treating lupus.
Sigma-Aldrich
GSK143 dihydrochloride, ≥97% (HPLC)