Merck
  • The VP3 structural protein of foot-and-mouth disease virus inhibits the IFN-β signaling pathway.
The VP3 structural protein of foot-and-mouth disease virus inhibits the IFN-β signaling pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2016-01-28)
Dan Li, Wenping Yang, Fan Yang, Huanan Liu, Zixiang Zhu, Kaiqi Lian, Caoqi Lei, Shu Li, Xiangtao Liu, Haixue Zheng, Hongbing Shu
ABSTRACT
Foot-and-mouth disease is a frequently occurring disease of cloven-hoofed animals that is caused by infection with the foot-and-mouth virus (FMDV). FMDV circumvents the type-I IFN response by expressing proteins that antagonize cellular innate immunity, such as leader protease and 3C protease. We identified the FMDV structural protein VP3 as a negative regulator of the virus-triggered IFN-β signaling pathway. Expression of FMDV VP3 inhibited the Sendai virus-triggered activation of IFN regulatory factor-3 and the expression of retinoic acid-inducible gene-I/melanoma differentiation-associated protein-5. Transient transfection and coimmunoprecipitation confirmed that the structural protein VP3 interacts with virus-induced signaling adapter (VISA), which is dependent on the C-terminal aa 111-220 of VP3. In addition, we found that FMDV VP3 inhibits the expression of VISA by disrupting its mRNA. Taken together, our findings reveal a novel strategy used by the structural VP3 protein of FMDV to evade host innate immunity.-Li, D., Yang, W., Yang, F., Liu, H., Zhu, Z., Lian, K., Lei, C., Li, S., Liu, X., Zheng, H., Shu, H. The VP3 structural protein of foot-and-mouth disease virus inhibits the IFN-β signaling pathway.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution