Merck
  • Repurposing drugs for treatment of tuberculosis: a role for non-steroidal anti-inflammatory drugs.
Repurposing drugs for treatment of tuberculosis: a role for non-steroidal anti-inflammatory drugs.
British medical bulletin (2016-05-07)
Arundhati Maitra, Sadé Bates, Monisha Shaik, Dimitrios Evangelopoulos, Ibrahim Abubakar, Timothy D McHugh, Marc Lipman, Sanjib Bhakta
ABSTRACT
The number of cases of drug-resistant Mycobacterium tuberculosis, the causative agent of tuberculosis (TB), has risen rapidly in recent years. This has led to the resurgence in repurposing existing drugs, such as non-steroidal anti-inflammatory drugs (NSAIDs), for anti-TB treatment. Evidence from novel drug screening in vitro, in vivo, pharmacokinetic/pharmacodynamics analyses and clinical trials has been used for the preparation of this systematic review of the potential of NSAIDs for use as an adjunct in new TB chemotherapies. Certain NSAIDs have demonstrated inhibitory properties towards actively replicating, dormant and drug-resistant clinical isolates of M. tuberculosis cells. NSAIDs are a diverse class of drugs, which have reported off-target activities, and their endogenous antimicrobial mechanism(s) of action is still unclear. It is essential that clinical trials of NSAIDs continue, in order to assess their suitability for addition to the current TB treatment regimen. Repurposing molecules such as NSAIDs is a vital, low-risk strategy to combat the trend of rapidly increasing antibiotic resistance.
Sigma-Aldrich
Carprofen, ≥97% (HPLC)