Merck
  • Glypican-1 identifies cancer exosomes and detects early pancreatic cancer.
Glypican-1 identifies cancer exosomes and detects early pancreatic cancer.
Nature (2015-06-25)
Sonia A Melo, Linda B Luecke, Christoph Kahlert, Agustin F Fernandez, Seth T Gammon, Judith Kaye, Valerie S LeBleu, Elizabeth A Mittendorf, Juergen Weitz, Nuh Rahbari, Christoph Reissfelder, Christian Pilarsky, Mario F Fraga, David Piwnica-Worms, Raghu Kalluri
ABSTRACT
Exosomes are lipid-bilayer-enclosed extracellular vesicles that contain proteins and nucleic acids. They are secreted by all cells and circulate in the blood. Specific detection and isolation of cancer-cell-derived exosomes in the circulation is currently lacking. Using mass spectrometry analyses, we identify a cell surface proteoglycan, glypican-1 (GPC1), specifically enriched on cancer-cell-derived exosomes. GPC1(+) circulating exosomes (crExos) were monitored and isolated using flow cytometry from the serum of patients and mice with cancer. GPC1(+) crExos were detected in the serum of patients with pancreatic cancer with absolute specificity and sensitivity, distinguishing healthy subjects and patients with a benign pancreatic disease from patients with early- and late-stage pancreatic cancer. Levels of GPC1(+) crExos correlate with tumour burden and the survival of pre- and post-surgical patients. GPC1(+) crExos from patients and from mice with spontaneous pancreatic tumours carry specific KRAS mutations, and reliably detect pancreatic intraepithelial lesions in mice despite negative signals by magnetic resonance imaging. GPC1(+) crExos may serve as a potential non-invasive diagnostic and screening tool to detect early stages of pancreatic cancer to facilitate possible curative surgical therapy.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture